Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD79b Hamster anti-Mouse, BUV395, Clone: HM79b (also known as HM79-16), BD Optibuild™

Catalog No. BDB740272 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740272 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740272 Supplier BD Biosciences Supplier No. 740272
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The HM79b monoclonal antibody specifically recognizes an extracellular epitope of Ig β chain (Ig β or CD79b), a 35-40-kDa transmembrane protein which forms an 80-90-kDa disulfide-linked heterodimer with Ig α chain (Ig α or CD79a, 30-35 kDa). On mature B lymphocytes, the CD79a/CD79b heterodimers are non-covalently associated with surface IgM to form the B-cell receptor complex (BCR). The presence of CD79a/CD79b is necessary for surface expression of the BCR and signal transduction via the BCR in B lymphocytes and pre-B cells. It was recently reported that CD79b may be expressed on the cell surface preceding the appearance of surface IgM during B-lymphocyte development. At this pro-B-cell stage, CD79b participates in signal transduction involved in the regulation of B-cell development. It should be noted that multi-parameter flow cytometric analyses of bone marrow suspensions performed at BD Biosciences Pharmingen have been unable to detect surface staining by HM79b mAb on CD45R/B220+ IgM- cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD79b
Applications Flow Cytometry
Classification Monoclonal
Clone HM79b (also known as HM79-16)
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Description Igβ; Igb; Ig-beta; Immunoglobulin-associated beta
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen Mouse B lymphoma, WEHI-123
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2 λ1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.