Learn More
CD80 Hamster anti-Mouse, Brilliant Violet 711, Clone: 16-10A1, BD Optibuild™
Hamster Monoclonal Antibody
Supplier: BD Biosciences 740698
Description
The 16-10A1 antibody specifically recognizes CD80 (B7-1). This member of the Ig superfamily, along with CD86 (B7-2), participates in T-cell co-stimulation via interactions with CD28 and CD152 (CTLA-4). CD80 has been reported to be constitutively expressed on dendritic cells, monocytes, and peritoneal macrophages; and it is inducible on B cells by various means, including activation by LPS, IL-4, and the cross-linking of surface Ig. Expression of CD80 has been reported to be greatly enhanced on splenic B cells following activation by LPS, with peak expression occurring between 48 and 72 hours. It has been reported that the activation of purified B cells with LPS can induce CD80 expression in as few as 18 hours. The 16-10A1 antibody has been reported to block binding of CTLA-4 Ig to CD80 and to block T-cell activation by Con A-elicited peritoneal exudate cells and CD80-transfected cell lines. However, 16-10A1 antibody alone is not able to block T-cell activation by antigen-presenting cells. CD86 (B7-2) is an alternate ligand for CD28 and CD152 (CTLA-4). Preliminary reports indicate that the 16-10A1 mAb may block the binding of rat anti-CD80 mAb clone 1G10 (Cat. No. 553368). In addition, it has been reported that the 16-10A1 antibody may cross-react with an activation antigen expressed on IFN- γ-activated alveolar macrophages of the dog.
The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.
Specifications
| CD80 | |
| Monoclonal | |
| 0.2mg/mL | |
| Cd80; B7; B7-1; Cd28l; Ly-53; MIC17; TSA1 | |
| Hamster | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2 κ |
| Flow Cytometry | |
| 16-10A1 | |
| Brilliant Violet 711 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse CD80 (B7) Transfected Cell Line | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only