Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ndfip1, Hamster, Clone: N1Nterm15-32.8, Abnova™

Catalog No. 89111012 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-012 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-012 Supplier Abnova Corporation Supplier No. MAB2460
Add to Cart
Add to Cart

Hamster monoclonal antibody raised against synthetic peptide of Ndfip1.

Sequence: VEPACGSGYQQLQNEEPGE

Specifications

Antigen Ndfip1
Applications ELISA, Western Blot
Classification Monoclonal
Clone N1Nterm15-32.8
Conjugate Unconjugated
Description Hamster monoclonal antibody raised against synthetic peptide of Ndfip1.
Dilution Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Ndfip1
Gene Alias 0610010M22Rik/N4wbp5
Gene Symbols Ndfip1
Host Species Hamster
Immunogen A synthetic peptide (conjugated with KLH) corresponding to mouse Ndfip1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65113
Test Specificity Mouse Ndfip1, N-terminus.
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.