Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49a Hamster anti-Mouse, Brilliant Violet 510, Clone: Ha31/8, BD Optibuild™

Catalog No. BDB740144 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740144 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740144 Supplier BD Biosciences Supplier No. 740144
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The Ha31/8 monoclonal antibody specifically binds to the 180-kDa integrin α 1 chain (CD49a), which is a transmembrane glycoprotein that non-covalently associates with the integrin β 1 subunit (CD29) to form the α 1β 1 (complex known as VLA-1). VLA-1 has been reported to be expressed on activated T cells, monocytes, smooth muscle cells, and endothelial cells. It is a receptor for collagen and laminin. The Ha31/8 monoclonal antibody is specific for both rat and mouse CD49a. It has been reported that Ha31/8 antibody can block VLA-1-mediated binding of rat cells to collagen.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49a
Applications Flow Cytometry
Classification Monoclonal
Clone Ha31/8
Concentration 0.2mg/mL
Conjugate Brilliant Violet 510
Description Itga1; Integrin alpha-1; Integrin α1; Laminin and collagen receptor; VLA-1a
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen Rat Emulsified Lewis Rat Glomeruli Cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2 λ1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.