Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpina3g, Hamster, Clone: MoFo29.2, Abnova™

Catalog No. 89111011 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-011 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-011 Supplier Abnova Corporation Supplier No. MAB2459
Add to Cart
Add to Cart

Hamster monoclonal antibody raised against synthetic peptide of Serpina3g.

Sequence: NPERSTNFPNGEGASSQR

Specifications

Antigen Serpina3g
Applications Western Blot
Classification Monoclonal
Clone MoFo29.2
Conjugate Unconjugated
Description Hamster monoclonal antibody raised against synthetic peptide of Serpina3g.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Serpina3g
Gene Alias 2A2/AI119734/MGC107057/Spi2-1/Spi2/eb.1/Spi2A/spi2
Gene Symbols Serpina3g
Host Species Hamster
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 406-426 of mouse Serpina3g.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20715
Test Specificity Mouse Serine protease inhibitor 2A.
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.