Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tcrb, Hamster, FITC, Clone: H57-597, Abnova™

Catalog No. 89112434 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-434 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-434 Supplier Abnova Corporation Supplier No. MAB5763
Add to Cart
Add to Cart

Hamster monoclonal antibody raised against native Tcrb.

Specifications

Antigen Tcrb
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone H57-597
Conjugate FITC
Description Hamster monoclonal antibody raised against native Tcrb.
Dilution Flow Cytometry (2 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Tcrb
Gene Alias Tib
Gene Symbols Tcrb
Host Species Hamster
Immunogen Native purified mouse Tcrb.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21577
Test Specificity Common epitope of the beta chain of the mouse alphabeta T cell receptor (TCR) complex.
Target Species Mouse
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.