Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tnfrsf8, Hamster, PE, Clone: 2SH12-5F, Abnova™

Catalog No. 89112384 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-384 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-384 Supplier Abnova Corporation Supplier No. MAB5711
Add to Cart
Add to Cart

Hamster monoclonal antibody raised against partial recombinant Tnfrsf8.

Specifications

Antigen Tnfrsf8
Applications Flow Cytometry
Classification Monoclonal
Clone 2SH12-5F
Conjugate PE
Description Hamster monoclonal antibody raised against partial recombinant Tnfrsf8.
Dilution Flow Cytometry (0.2 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Tnfrsf8
Gene Alias Cd30/D1S166E/Ki/Ki-1
Gene Symbols Tnfrsf8
Host Species Hamster
Immunogen Recombinant mouse IgG1 fusion protein corresponding to extracellular region of mouse Tnfrsf8.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21941
Test Specificity Murine CD30, Mr 52 KDa (core peptide).
Target Species Mouse
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.