Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Hand Centrifuge, 2 Bucket
SCI_ED

Catalog No. S40584 Shop All Eisco Products
Change view
Click to view available options
Capacity:
2 x 15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
S40584 2 x 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S40584 Supplier Eisco™ Supplier No. CH0266A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Cetrifuge, Screw clamp, Removable crank handle, 2 plastic buckets,

This hand driven centrifuge can accommodate two 15 ml tubes. Made with die cast parts, a steel pinion, and worm drive. Designed as an economical alternative to electrical centrifuges, this centrifuge must be attached to a lab bench via the screw clamp when in use. Crank handle can be removed.

  • Economical hand driven centrifuge
  • Accommodates two 15 ml tubes
  • Includes die cast parts, steel pinion, worm drive, 2 plastic buckets
  • Attaches to lab bench or table via screw clamp (included)
  • Removable crank handle for easy storage

Warranty and Services

1 year limited manufacturer warranty

Specifications

Capacity 2 x 15 mL
Max. Speed Not Applicable
Max. RCF Not Applicable
Includes 4 buckets, hand crank, screw clamp
For Use With (Equipment) 15 mL tubes
Refrigerated No
Color Gray
Quantity 1 Ea.
Product Type Manual
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.