Learn More
Invitrogen™ HAND1 Monoclonal Antibody (OTI1G10)

Description
HAND1 Monoclonal Antibody for Western Blot, ICC/IF, Flow
Specifications
Specifications
| Antigen | HAND1 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI1G10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | HAND1 |
| Gene Accession No. | O96004 |
| Gene Alias | bHLHa27; Class A basic helix-loop-helix protein 27; eHAND; Ehand1; extraembryonic tissues, heart, autonomic nervous system and neural crest derivatives-expressed protein 1; HAND1; heart and neural crest derivatives expressed 1; heart and neural crest derivatives expressed transcript 1; heart- and neural crest derivatives-expressed protein 1; Helix-loop-helix transcription factor expressed in extraembryonic mesoderm and trophoblast; Hxt; Th1; Thing1; thing-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.