Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HAND2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115339
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115339 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115339 Supplier Invitrogen™ Supplier No. PA5115339
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HAND2.

The protein encoded by this gene belongs to the basic helix-loop-helix family of transcription factors. This gene product is one of two closely related family members, the HAND proteins, which are asymmetrically expressed in the developing ventricular chambers and play an essential role in cardiac morphogenesis. Working in a complementary fashion, they function in the formation of the right ventricle and aortic arch arteries, implicating them as mediators of congenital heart disease. In addition, this transcription factor plays an important role in limb and branchial arch development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAND2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene HAND2
Gene Accession No. P61295, P61296, Q61039
Gene Alias AI225906; AI661148; basic helix-loop-helix transcription factor HAND2; bHLHa26; Class A basic helix-loop-helix protein 26; deciduum, heart, autonomic nervous system and neural crest derivatives-expressed protein 2; DHAND; dHAND basic helix-loop-helix transcription factor; dHand protein; DHAND2; Ehand2; han; Hand2; hands off; heart and neural crest derivatives expressed 2; heart and neural crest derivatives expressed transcript 2; heart- and neural crest derivatives-expressed protein 2; Hed; helix-loop-helix transcription factor expressed in embryo and deciduum; Th2; Thing2; thing-2; zdHAND; zgc:103415
Gene Symbols HAND2
Host Species Rabbit
Immunogen A synthesized peptide derived from human HAND2(Accession P61296), corresponding to amino acid residues G45-P95.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15111, 64637, 9464
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.