Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hantaan Virus Nucleocapsid Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
| Antigen | Hantaan Virus Nucleocapsid |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Hantaan Virus Nucleocapsid Polyclonal specifically detects Hantaan Virus Nucleocapsid in Virus samples. It is validated for ELISA.Specifications
| Hantaan Virus Nucleocapsid | |
| ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| Antibody was raised against an 18 amino acid peptide near the amino terminus of the Hantaan virus nucleocapsid. The immunogen is located within the first 50 amino acids of Hantaan Virus Nucleocapsid. Amino Acid Squence: YEKDPDELNKRTLTDREG | |
| Primary | |
| 48 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Virus | |
| HTNVsSgp1 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title