Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Hantavirus NP Monoclonal Antibody (Q11S)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA518208
Description
This antibody is specific for the nucleocapsid protein (N) of all Hantaviruses tested: Hantaan, Sin Nombre, Puumala, Dobrava, Seoul and Saaremaa. Purity is >90%.

Specifications
Hantavirus NP | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry | |
Q11S | |
Unconjugated | |
HV; HV nucleoprotein | |
Hantavirus recombinant protein. | |
100 μg | |
Primary | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction