Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HAO1 Polyclonal Antibody
Rabbit Polyclonal Antibody
$237.00 - $511.50
Specifications
Antigen | HAO1 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119199
|
Invitrogen™
PA5119199 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121928
|
Invitrogen™
PA5121928 |
20 μL |
Each for $237.00
|
|
|||||
Description
HAO1 Polyclonal Antibody for Western Blot, IHC (P)
Has 2-hydroxyacid oxidase activity. Most active on the 2-carbon substrate glycolate, but is also active on 2-hydroxy fatty acids, with high activity towards 2-hydroxy palmitate and 2-hydroxy octanoate.Specifications
HAO1 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Mouse, Rat, Pig | |
HAO1 | |
(S)-2-hydroxy-acid oxidase; glycolate oxidase; glycolate oxidase 1; GOX; Gox1; GOX1 antibody; Hao1; Hao-1; HAOX1; HAOX1 antibody; hydroxyacid oxidase (glycolate oxidase) 1; hydroxyacid oxidase 1; hydroxyacid oxidase 1, liver; MGC142225 antibody; XDH1 | |
HAO1 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q9UJM8, Q9WU19 | |
100627803, 15112, 311446, 54363 | |
Recombinant protein Hydroxyacid Oxidase 1. The antigen corresponds to amino acid range 113-369 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title