Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HAO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA593000
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA593000 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA593000 Supplier Invitrogen™ Supplier No. PA593000
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLPRLICIND YEQHAKSVLP KSIYDYYRSG ANDEETLADN IAAFSRWKLY PRMLRNVAET DLSTSVLGQR VSMPICVGAT AMQRMAHVDG ELATVRACQS LGTGMMLSSW ATSSIEEVAE AGPEALRWLQ LYIYKDREVT KKLVRQAEKM GYKAIFVTVD TPYLGNRLDD VRNRFKLPPQ LRMKNFETST LSFSPEENFG DDSGLAAYVA KAIDPSISWE DIKWLRRLTS LPIVAKGILR GDDAREAVKH GLNGILVSNH GARQLDGVPA TIDVLPEIVE AVEGKVEVFL DGGVRKGTDV LKALALGAKA VFVGRPIVWG LAFQGEKGVQ DVLEILKEEF RLAMALSGCQ NVKVIDKTLV RKNPLAVSKI; Positive Samples: Mouse liver, Rat liver; Cellular Location: Peroxisome.

Has 2-hydroxyacid oxidase activity. Most active on the 2-carbon substrate glycolate, but is also active on 2-hydroxy fatty acids, with high activity towards 2-hydroxy palmitate and 2-hydroxy octanoate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAO1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HAO1
Gene Accession No. Q9UJM8, Q9WU19
Gene Alias (S)-2-hydroxy-acid oxidase; glycolate oxidase; glycolate oxidase 1; GOX; Gox1; GOX1 antibody; Hao1; Hao-1; HAOX1; HAOX1 antibody; hydroxyacid oxidase (glycolate oxidase) 1; hydroxyacid oxidase 1; hydroxyacid oxidase 1, liver; MGC142225 antibody; XDH1
Gene Symbols HAO1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-370 of human HAO1/GOX (NP_0600151).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15112, 311446, 54363
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.