Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAO1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988674
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8674 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8674 Supplier Bioss Supplier No. BS8602R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Has 2-hydroxyacid oxidase activity. Most active on the 2-carbon substrate glycolate, but is also active on 2-hydroxy fatty acids, with high activity towards 2-hydroxy palmitate and 2-hydroxy octanoate.

Specifications

Antigen HAO1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HAO1
Gene Accession No. Q9UJM8
Gene Alias (S)-2-hydroxy-acid oxidase; Glycolate oxidase; glycolate oxidase 1; GOX; Gox1; GOX1 antibody; Hao1; Hao-1; HAOX1; HAOX1 antibody; hydroxyacid oxidase (glycolate oxidase) 1; Hydroxyacid oxidase 1; hydroxyacid oxidase 1, liver; MGC142225 antibody; XDH1
Gene Symbols HAO1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GOX/HAO1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54363
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.