Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HAO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590263
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590263 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590263 Supplier Invitrogen™ Supplier No. PA590263
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSLVCLTDFQ AHAREQLSKS TRDFIEGGAD DSITRDDNIA AFKRIRLRPR YLRDVSEVDT RTTIQGEEIS APICIAPTGF HCLVWPDGEM STARAAQAAG ICYITSTFAS CSLEDIVIAA PEGLRWFQLY VHPDLQLNKQ LIQRVESLGF KALVITLDTP VCGNRRHDIR NQLRRNLTLT DLQSPKKGNA IPYFQMTPIS TSLCWNDLSW FQSITRLPII LKGILTKEDA ELAVKHNVQG IIVSNHGGRQ LDEVLASIDA LTEVVAAVKG KIEVYLDGGV RTGNDVLKAL ALGAKCIFLG RPILWGLACK GEHGVKEVLN ILTNEFHTSM ALTGCRSVAE INRNLVQFSR L; Positive Samples: Mouse kidney; Cellular Location: Peroxisome.

HAO2 gene ontology annotations related to this gene include fatty acid oxidation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAO2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene HAO2
Gene Accession No. Q9NYQ2
Gene Alias (S)-2-hydroxy-acid oxidase, peroxisomal; (S)-hydroxy-acid oxidase, peroxisomal; AI325478; Cell growth-inhibiting gene 16 protein; GIG16; glycolate oxidase; Hao2; Hao-2; Hao3; HAOX2; Haox3; hydroxyacid oxidase (glycolate oxidase) 3; hydroxyacid oxidase 2; hydroxyacid oxidase 2 (long chain); hydroxyacid oxidase 2, kidney; hydroxyacid oxidase 3 (medium-chain); hypothetical protein LOC509481; long chain alpha-hydroxy acid oxidase; Long-chain L-2-hydroxy acid oxidase; medium chain alpha-hydroxy acid oxidase; medium-chain 2-hydroxy acid oxidase HAOX3; medium-chain L-2-hydroxy acid oxidase
Gene Symbols HAO2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-351 of human HAO2 (NP_0576111).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56185
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.