Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAP1 Antibody (1B6), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 8 publications

$192.47 - $517.59

Specifications

Antigen HAP1
Clone 1B6
Concentration 1 mg/mL
Dilution Western Blot 0.5-1.0 ug/ml, Flow Cytometry 1 ug per million cells, Immunohistochemistry 0.5-1.0 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 0.5-1.0 ug/ml, Immunohistochemistry-Frozen 0.5-1.0 ug/ml
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11074569T
SDP
View Documents
Novus Biologicals
NB11074569SS
0.025 mL
Each for $192.47
Only null left
Add to Cart
 
NB11074569
SDP
View Documents
Novus Biologicals
NB11074569
0.1 mL
Each for $517.59
Only null left
Add to Cart
 
Description

Description

HAP1 Monoclonal specifically detects HAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

HAP1
1 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Mouse
Autophagy, Neuroscience
HAP2huntingtin-associated protein 2, hHLP1, HIP5, HLP1, HLPHAP-1, huntingtin-associated protein 1, Neuroan 1
HAP1
IgG1
Protein G purified
This is specific for isoforms A and B.
1B6
Western Blot 0.5-1.0 ug/ml, Flow Cytometry 1 ug per million cells, Immunohistochemistry 0.5-1.0 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 0.5-1.0 ug/ml, Immunohistochemistry-Frozen 0.5-1.0 ug/ml
Monoclonal
Purified
RUO
Human, Mouse, Rat
9001
Fusion protein from rat HAP1. [UniProt# P54256]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.