Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HAP1 Antibody (1B6), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11074569PEATT594
Description
HAP1 Monoclonal specifically detects HAP1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
HAP1 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
HAP1 | |
Fusion protein from rat HAP1. [UniProt No. P54256] | |
0.1 mL | |
Primary | |
This is specific for isoforms A and B. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
1B6 | |
Flow Cytometry | |
HAP2huntingtin-associated protein 2, hHLP1, HIP5, HLP1, HLPHAP-1, huntingtin-associated protein 1, Neuroan 1 | |
Mouse | |
Protein G purified | |
Autophagy, Neuroscience | |
9001.0 | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction