Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAP1, Mouse anti-Mouse, Rat, Clone: 1B6, Millipore Sigma™

Catalog No. MABN2456100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2456100 100 μL
MABN245625 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN2456100 Supplier MilliporeSigma Supplier No. MABN2456100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-HAP1, clone 1B6, Cat. No. MABN2456, is a highly specific mouse monoclonal antibody that targets Huntingtin-associated protein 1 and has been tested for use in Immunocytochemistry, Immunoprecipitation, and Western Blotting.

Huntingtin-associated protein 1 (UniProt: P54256; also known as HAP1) is encoded by the Hap1 gene (Gene ID: 29430) in rat. HAP1 is a huntingtin-associated protein that is enriched in the brain. It specifically associates with HTT/huntingtin and this binding is enhanced by an expanded polyglutamine repeats within HTT that affects HAP1 interaction properties. Both HTT and HAP1 are reported to be involved in intracellular trafficking and HAP1 is proposed to link HTT to motor proteins and/or transport cargos. HAP1 localizes to large nonmembrane-bound cytoplasmic bodies (stigmoid bodies) in various types of neurons. The formation of stigmoid bodies is regulated by the ratio of isoform A to isoform B of HAP1. HAP1 plays a role in vesicular transport within neurons that involves microtubule-associated transporters and is shown to be attenuated by association with mutant HTT. In rodents, two HAP1 isoforms (HAP1A and HAP1B) have been described that differ in their carboxy terminal. However, humans contain only the HAP1A isoform. In murine models, targeted disruption of the HAP1 gene results in significantly depressed feeding behavior and premature postnatal death. Pups with Hap1 -/- show decreased feeding, malnutrition, dehydration, and premature death. (Ref.: Tang, TS et al., (2003). Neuron 39 (2); 227-239; Chan, EY., et al. (2002). Hum. Mol. Gen. 11(8); 945-959).
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAP1
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1B6
Conjugate Unconjugated
Dilution Immunocytochemistry Analysis: A representative lot detected HAP1 in Immunocytochemistry applications (Fujinaga, R., et. al. (2011). Exp. Cell Res. 317(12):1689-700).Immunoprecipitation Analysis: A representative lot immunoprecipitated HAP1 in Immunoprecipitation applications (Fujinaga, R., et. al. (2011). Exp. Cell Res. 317(12):1689-700).Western Blotting Analysis: A representative lot detected HAP1 in Western Blotting applications (Chan, E.Y., et. al (2002). Hum Mol Genet. 11(8):945-59; Tang, T.S., et. al. (2003). Neuron. 39(2):227-39; Torre, E.R., et. al. (2003). J Neurosci Methods. 125(1-2):27-32).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Huntingtin-associated protein 1
Gene Symbols Hap1
Host Species Mouse
Immunogen GST-tagged recombinant fragment corresponding to 137 amino acids from the internal region of rat Huntingtin-associated protein 1 (HAP1).
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_077047.1
Test Specificity Clone 1B6 detects both HAP1A and HAP1B proteins in mouse and rat brain. It targets an epitope from the internal region.
Target Species Mouse, Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.