Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HAP40 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP302964100UL
Description
HAP40 Polyclonal specifically detects HAP40 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
HAP40 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
Unconjugated | |
coagulation factor VIII-associated (intronic transcript) 1, coagulation factor VIII-associated 1, cpG island protein, DXS522E, F8A, factor VIII associated protein, factor VIII intron 22 protein, HAP40, huntingtin-associated protein 40 | |
A synthetic peptide corresponding to a sequence within amino acids 150-250 of humanHAP40 (NP_036283.2). | |
100 μL | |
Autophagy, Neuronal Cell Markers, Neuroscience | |
8263 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction