Learn More
Invitrogen™ HAPLN1 Polyclonal Antibody

Description
HAPLN1 Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | HAPLN1 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | HAPLN1 |
| Gene Accession No. | P10915 |
| Gene Alias | BB099155; Cartilage link protein; cartilage link protein 1; cartilage linking protein 1; cartilage-link protein; Cartilage-linking protein 1; CLP; CRT1; Crtl1; Crtl1l; fc26f11; fc37g04; fc50e12; fc55h01; HAPLN1; hapln1a; Hyaluronan and proteoglycan link protein 1; hyaluronan and proteoglycan link protein 1a; hyaluronan proteoglycan link protein 1; link protein; LP; LP-1; Proteoglycan link protein; wu:fc26f11; wu:fc37g04; wu:fc50e12; wu:fc55h01 |
| Gene Symbols | HAPLN1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.