Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HAPLN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA593098
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA593098 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA593098 Supplier Invitrogen™ Supplier No. PA593098
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PRLGRYNLNF HEAQQACLDQ DAVIASFDQL YDAWRGGLDW CNAGWLSDGS VQYPITKPRE PCGGQNTVPG VRNYGFWDKD KSRYDVFCFT SNFNGRFYYL IHPTKLTYDE AVQACLNDGA QIAKVGQIFA AWKILGYDRC DAGWLADGSV RYPISRPRRR CSPTEAAVRF VGFPDKKHKL YGVYCFRAYN; Positive Samples: HeLa, A375, Mouse liver; Cellular Location: extracellular matrix, extracellular space.

HAPLN1 belongs to the HAPLN family and contains 1 Ig like V type (immunoglobulin like) domain and 2 Link domains. HAPLN1 stabilizes the aggregates of proteoglycan monomers with hyaluronic acid in the extracellular cartilage matrix. Diseases associated with HAPLN1 include Cerebral Creatine Deficiency Syndrome 3 and Rheumatoid Arthritis, Systemic Juvenile.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAPLN1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene HAPLN1
Gene Accession No. P10915, Q9QUP5
Gene Alias BB099155; Cartilage link protein; cartilage link protein 1; cartilage linking protein 1; cartilage-link protein; Cartilage-linking protein 1; CLP; CRT1; Crtl1; Crtl1l; fc26f11; fc37g04; fc50e12; fc55h01; HAPLN1; hapln1a; Hyaluronan and proteoglycan link protein 1; hyaluronan and proteoglycan link protein 1a; hyaluronan proteoglycan link protein 1; link protein; LP; LP-1; Proteoglycan link protein; wu:fc26f11; wu:fc37g04; wu:fc50e12; wu:fc55h01
Gene Symbols HAPLN1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 165-354 of human HAPLN1 (NP_0018751).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12950, 1404
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.