Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HAPLN1 Rabbit anti-Human, Mouse, Rat, Clone: 5O9U2, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP31648620UL
This item is not returnable.
View return policy
Description
HAPLN1 Monoclonal antibody specifically detects HAPLN1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
HAPLN1 | |
Monoclonal | |
Unconjugated | |
Cartilage link protein, Cartilage-link protein, Cartilage-linking protein 1, CRTL1cartilage linking protein 1, hyaluronan and proteoglycan link protein 1, Proteoglycan link protein | |
A synthetic peptide corresponding to a sequence within amino acids 250-354 of human HAPLN1 (P10915). | |
20 μg | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
1404 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
5O9U2 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction