Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HAPLN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520497
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520497 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520497 Supplier Invitrogen™ Supplier No. PA520497
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human brain tissue lysate. PA5-20497 can be used with blocking peptide PEP-0617.

BRAL1 is a member a superfamily consisting of several highly homologous hyaluronan and proteoglycan binding link proteins. BRAL1 is predominantly expressed in brain tissue and spinal cord. Like other members in the link-module superfamily, BRAL1 contains an immunoglobulin-like fold and two proteoglycan tandem repeats (PTRs). Its mRNA expression pattern is similar to other lectican proteoglycans, suggesting that BRAL1 may act to stabilize the binding between the extracellular matrix molecule hyaluronan and brevican. Immunostaining of mouse brain showed BRAL1 expression at P20 in the white matter of the developing cerebellum and in myelinated fiber tracts in the adult brain, suggesting that expression starts when axonal myelination occurs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAPLN2
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene HAPLN2
Gene Accession No. Q9ESM2, Q9ESM3, Q9GZV7
Gene Alias 4930401E20Rik; Brain link protein 1; brain link protein-1; Bral1; HAPLN2; Hyaluronan and proteoglycan link protein 2; RP11-284F21.6
Gene Symbols HAPLN2
Host Species Rabbit
Immunogen a 12 amino acid peptide from near the center of human BRAL1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60484, 64057, 73940
Target Species Human, Mouse, Rat
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.