Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HAPLN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558708
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558708 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558708 Supplier Invitrogen™ Supplier No. PA558708
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GLPFYNGFYY SNSANDQNLG NGHGKDLLNG VKLVVETPEE TLFTYQGASV ILPCRYRYEP ALVSPRRVRV KWWKLSEN Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 74%.

HAPLN3 is a 360 amino acid extracellular protein belonging to the HAPLN family and contains one Ig-like V-type (immunoglobulin-like) domain and two Link domains. This protein may function in hyaluronic acid binding and cell adhesion. It is either a secreted or an extracellular matrix protein involved in stabilizing the aggregates of proteoglycan monomers with hyaluronic acid in the extracellular cartilage matrix. HAPLN3 is widely expressed in most tissues with highest levels in spleen and placenta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAPLN3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HAPLN3
Gene Accession No. Q96S86
Gene Alias 4930554N11Rik; EXLD1; extracellular link domain containing, 1; Hapln3; HsT19883; Hyaluronan and proteoglycan link protein 3; link protein 3; Lpr3; UNQ238/PRO271
Gene Symbols HAPLN3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HAPLN3. Recombinant protein control fragment (Product #RP-98111).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145864
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.