Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HAPLN4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547574
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547574 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547574 Supplier Invitrogen™ Supplier No. PA547574
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In Western blots, less than 1% cross-reactivity with recombinant human HAPLN1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Hyaluronan and proteoglycan link protein 4 (HAPLN4), also known as Brain Link Protein 4 (BRAL1), is a gene located on chromosome 5q14.2, encoding a member of the link protein family that plays a crucial role in stabilizing interactions between hyaluronan and proteoglycans within the extracellular matrix. HAPLN4 is predominantly expressed in the brain, especially within the central nervous system where it contributes to the structural integrity and functional modulation of neural tissues. The protein is key in maintaining the organization of the extracellular matrix by binding hyaluronan and other matrix components, facilitating cell adhesion and signaling. This is vital for processes like neuronal migration, synapse formation, and neuronal differentiation during brain development. Variations in HAPLN4 expression or function are associated with neural developmental disorders and may influence the progression of neurodegenerative diseases due to its critical role in preserving the extracellular matrix architecture and neural connectivity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAPLN4
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene HAPLN4
Gene Accession No. Q86UW8
Gene Alias 9330174O11; Brain link protein 2; brain linked protein 2; Bral2; Hapln4; Hyaluronan and proteoglycan link protein 4; hyaluronan and proteoglycan link protein 4; LOW QUALITY PROTEIN: hyaluronan and proteoglycan link protein 4; KIAA1926; link protein 4; LOW QUALITY PROTEIN: hyaluronan and proteoglycan link protein 4; Lpr4; si:dkey-19f23.2
Gene Symbols HAPLN4
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human HAPLN4 Gln30-Val402.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 404037
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.