Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAPLN4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983743
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3743 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3743 Supplier Bioss Supplier No. BS15412R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HAPLN4 binds to hyaluronic acid and may be involved in formation of the extracellular matrix.

Specifications

Antigen HAPLN4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HAPLN4
Gene Accession No. Q86UW8
Gene Alias 9330174O11; Brain link protein 2; brain linked protein 2; Bral2; HAPLN4; Hyaluronan and proteoglycan link protein 4; KIAA1926; link protein 4; LOW QUALITY PROTEIN: hyaluronan and proteoglycan link protein 4; Lpr4; si:dkey-19f23.2
Gene Symbols HAPLN4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HAPLN4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 404037
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.