Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hapten 4-hydroxy-3-nitrophenyl acetyl (NP) Antibody (B1-8), Janelia Fluor™ 549 - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP252640JF549
Description
hapten 4-hydroxy-3-nitrophenyl acetyl (NP) Monoclonal specifically detects hapten 4-hydroxy-3-nitrophenyl acetyl (NP) in Non-species specific samples. It is validated for Western Blot, Flow Cytometry.Specifications
hapten4-hydroxy-3-nitrophenylacetyl(NP) | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Rabbit | |
Protein A or G purified | |
Primary | |
Non-species specific | |
Purified |
Flow Cytometry | |
B1-8 | |
Flow Cytometry | |
(4-Hydroxy-3-nitrophenyl)acetyl, 2-(4-Hydroxy-3-nitrophenyl)acetate, 2-(4-Hydroxy-3-nitrophenyl)acetic acid, 3-nitro-4-Hydroxyphenylacetic acid, 4-Hydroxy-3-nitrophenyl acetyl | |
hapten-protein conjugated NP12-chicken gammaglobulin (CG). | |
0.1 mL | |
Hapten (4-hydroxy-3-nitropheny1) acetyl (NP). | |
Store at 4°C in the dark. | |
IgG λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction