Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hapten 4-hydroxy-3-nitrophenyl acetyl (NP) Antibody (B1-8), mFluor Violet 500 SE, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP281027MFV500
Description
hapten 4-hydroxy-3-nitrophenyl acetyl (NP) Monoclonal antibody specifically detects hapten 4-hydroxy-3-nitrophenyl acetyl (NP) in Non-species specific samples. It is validated for Flow CytometrySpecifications
hapten 4-hydroxy-3-nitrophenyl acetyl (NP) | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Human | |
Protein A purified | |
RUO | |
Non-species specific | |
Purified |
Flow Cytometry | |
B1-8 | |
Flow Cytometry | |
(4-Hydroxy-3-nitrophenyl)acetyl, 2-(4-Hydroxy-3-nitrophenyl)acetate, 2-(4-Hydroxy-3-nitrophenyl)acetic acid, 3-nitro-4-Hydroxyphenylacetic acid, 4-Hydroxy-3-nitrophenyl acetyl | |
hapten-protein conjugated NP12-chicken gammaglobulin (CG). | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction