Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hapten 4-hydroxy-3-nitrophenyl acetyl NP Chimeric Recombinant Rabbit Monoclonal Antibody (B1-8)
GREENER_CHOICE

Catalog No. PIMA547758
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547758 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547758 Supplier Invitrogen™ Supplier No. MA547758
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: Hapten (4-hydroxy-3-nitropheny1) acetyl (NP).

Hapten 4-hydroxy-3-nitrophenyl acetyl NP is a small molecule that consists of a hapten, which is the 4-hydroxy-3-nitrophenyl acetyl group, linked to a carrier protein, which is NP or nitrophenol. The carrier protein is an important component of this molecule as it helps to elicit an immune response by presenting the hapten portion of the molecule to the immune system. NP is commonly used as a carrier in immunology research because it is a small, soluble molecule that is easily conjugated with haptens. When the hapten binds to specific antibodies, the carrier protein helps to stimulate an immune response and activate B cells, which can lead to the production of specific antibodies against the hapten.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hapten 4-hydroxy-3-nitrophenyl acetyl NP Chimeric
Applications Western Blot
Classification Recombinant Monoclonal
Clone B1-8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Host Species Rabbit
Immunogen hapten-protein conjugated NP12-chicken gammaglobulin (CG).
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG λ
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.