Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Haptoglobin Antibody (5A5), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Haptoglobin |
---|---|
Clone | 5A5 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
Haptoglobin Monoclonal antibody specifically detects Haptoglobin in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
Haptoglobin | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
binding peptide, BP, haptoglobin, haptoglobin alpha(1S)-beta, haptoglobin alpha(2FS)-beta, haptoglobin, alpha polypeptide, haptoglobin, beta polypeptide, HP2ALPHA2, HPA1S, MGC111141, zonulin | |
A synthesized peptide derived from Human Haptoglobin [UniProt P00738] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
5A5 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cardiovascular Biology | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
3240 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title