Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Haptoglobin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586629
Description
Purity is > 95% by SDS-PAGE.
Haptoglobin is an a2-globulin plasma protein synthesized in the liver. It is an acute phase protein which also acts as a scavenger of hemoglobin from hemolyzed red cells. The haptoglobin monomer consists of two a and two beta chains; the a chains bay be hp1Fa, hp1Sa, or hp2a types while the beta chain is constant. Three major phenotypes are recognized: HP1-1, Hp2-1 and Hp-2-2, in which the hp1a chain bay be either the F or S type.
Specifications
Haptoglobin | |
Polyclonal | |
Unconjugated | |
HP | |
alpha polypeptide; Ba1-647; beta polypeptide; binding peptide; Binding peptide (BP); BP; haptoglobin; haptoglobin alpha 1S; Haptoglobin alpha chain; Haptoglobin alpha chain / alpha(1S) beta; haptoglobin alpha(1S)-beta; Haptoglobin alpha(2FS) beta; haptoglobin alpha(2FS)-beta; Haptoglobin beta chain; haptoglobin, alpha polypeptide; haptoglobin, beta polypeptide; haptoglobin; haptoglobin-like; haptoglobin-like; haptoglobin-related protein; Hp; HP-1; HP2 ALPHA2; HP2ALPHA2; HP2-ALPHA-2; HPA1S; HPR; HPT; Liver regeneration-related protein LRRG173; LOC100067869; LOC479668; MGC111141; MGC133752; preHP2; zonulin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15439, 24464, 3240 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P00738, P06866, Q61646 | |
HP | |
Synthetic peptide, corresponding to amino acids 301-350 of Human Haptoglobin. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction