Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Westcott™ Hard Handle Kids Scissors
SCI_ED

Catalog No. S81746
Click to view available options
Description:
Blunt
Pointed

Products are specifically designed for growing students, created to be fun and functional with superior quality and exceptional value.

  • Especially designed for a child's hand.
  • Extra strong and durable stainless steel safety blades
  • Molded, asymmetrical handles allow for left handed use
  • Pointed scissors are most suitable for ages 4+
  • Easily cut through paper, string and other craft materials
  • Available in assorted colors (color can't be specified when ordered)
  • Manufacturer's limited five year warranty

Specifications

Product Type Scissor
Color Assorted
Description Pointed
Grade Pre-K to 12
Age 4 to 18
Length (English) 5 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.