Learn More
John M. Maris Company Hard Rubber Spatulas
Nonsparking, wide, flexible blade
$32.49 - $34.58
Specifications
Sterility Sterile
Product Type Spatula
For Use With (Application) Designed to used with materials that affect Metals adversely, for mixing, weighing, cleaning, blending and many other uses
Material Hard Rubber
Handle Type Integral
View More Specs
Products 3
Catalog Number Mfr. No. Length (Metric) Length (Metric) Blade Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Length (Metric) Blade Price Quantity & Availability  
14-355B
Encompass
View Documents
John M. Maris Company
106
26 cm 15.2 cm
Each for $33.54
Pack of 12 EA for $361.94
Only null left
Add to Cart
14-355A
Encompass
View Documents
John M. Maris Company
104
19 cm 10.2 cm
Each for $32.49
Pack of 12 EA for $336.36
Only null left
Add to Cart
14-355C
Encompass
View Documents
John M. Maris Company
108
33 cm 20.3 cm
Each for $34.58
Pack of 12 EA for $387.40
Only null left
Add to Cart
Description
Description
Nonsparking, wide, flexible blade
  • Integral handle
  • Can be used with materials that affect metals adversely
  • Washable
  • Sterilizable
Specifications
Specifications
Sterile
Designed to used with materials that affect Metals adversely, for mixing, weighing, cleaning, blending and many other uses
Integral
Spatula
Hard Rubber
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.