Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Hardness Kit
SDPSCI_ED

Catalog No. s98238
Click to view available options
:
Sensitivity: 4ppm CaCO3; No. of tests: 50 to 200
This set contains chemicals that may be harmful if misused. Read cautions on individual containers carefully. Not to be used by children except under adult supervision.
This set contains chemicals that may be harmful if misused. Read cautions on individual containers carefully. Not to be used by children except under adult supervision.

Includes

Hardness reagent no. 5, 15mL; hardness reagent no. 6 tablets, 100; hardness reagent no. 7, 60mL; titration test tube (5, 10, 12.9, 15, 20, and 25mL), glass, w/cap, 1 each; direct reading titrator (0 to 200 range), 1 each; 0.5mL plastic pipet

EDTA titration is used for all hardness determinations.

  • Liquid buffer and tablet indicator
  • Red to blue endpoint
  • Total hardness buffer includes inhibitors to eliminate metal interferences
  • All results are as CaCO3
  • Kit also expresses results as gpg

Specifications

Product Type Hardness Kit
For Use With (Application) For hardness determinations
Includes Hardness reagent no. 5, 15ml, hardness reagent no. 6 tablets, 100, hardness reagent no. 7, 60ml, titration test tube (5, 10, 12.9, 15, 20, and 25ml), glass, with cap, direct reading titrator (0 to 200 range), 0.5ml plastic pipet
Class Size 1 Class
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.