Learn More
StatLab™ Harris Hematoxylin Stain
GSA_VASDP
Catalog No. p-7227716
Click to view available options
Quantity:
4 x 1 gal.
4 x 16 oz. Bottle
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Modified traditional formulation of Harris Hematoxylin.
  • Contains no mercury
  • Generally used as a regressive stain, but may also be used in progressive staining protocols
  • Produces rapid and distinct nuclear detail
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Nuclear Stain
Quantity 4 x 16 oz. Bottle
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.