Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harris Industries Barricade Tapes

Catalog No. 19811825
Encompass
Click to view available options
Color:
Red
Yellow
Dimensions (L x W):
1000 ft. x 3 in.
300 ft. x 3 in.
Displayed Text:
Caution Caution Caution
Caution Construction Area
Caution Do Not Enter
Danger Danger
Danger Do Not Enter
Danger Hazardous Area Keep Out
Danger Hazardous Material Do Not Enter
Danger Keep Out
Fire Line Do Not Cross

Increase visibility of hazards to help prevent accidents

Strong, lightweight polyethylene tapes with continuous repetition of message.

  • Can be tied, stapled, or nailed into place
  • Economical and reusable
  • Caution tapes have yellow background
  • Danger tapes have red background

Order Info

Roll size: 3 in. W x 1000 ft.L (7.6cm x 304.8m). Thickness: 4mil.

Compliance

Meet OSHA 1910.144 for marking physical hazards.

Specifications

Dimensions (L x W) 1000 ft. x 3 in.
Thickness 4 mil
Material HDPE
Color Yellow
Certifications/Compliance OSHA 1910.144
Width (English) 3 in.
Width (Metric) 7.62 cm
Displayed Text Fire Line Do Not Cross
For Use With (Application) Increase visibility of hazards to help prevent accidents
Length (English) 1000 ft.
Length (Metric) 304.8 m
Type Municipal Tape
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.