Harris Industries Barricade Tapes

Increase visibility of hazards to help prevent accidents

Manufacturer: HARRIS INDUSTRIES INC BT052

 View more versions of this product

Catalog No. 19-811-821

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Length (English) 1000 ft.
    For Use With (Application) Increase visibility of hazards to help prevent accidents
    Width (English) 3 in.
    Width (Metric) 7.62cm
    Certification Compliance OSHA 1910.144
    Color Yellow
    Displayed Text Caution Caution Caution
    Material Polyethylene
    Product Type Caution Tape
    Dimensions (L x W) 200 ft. x 3 in. (60.9m x 7.62cm)
    Length (Metric) 304.8m
    Thickness 4mil

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit