Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Harris Industries Flagging Tape

Click to view available options
Color:
Blue
Orange
Pink
White
Yellow
Length (English):
150 ft.
300 ft.
Thickness:
2 mil
4 mil
Description
- 3cm (1.19 in.) width
- 3.81cm (1.5 in.) core
- Non-adhesive
- Vinyl with monomeric flexible PVC film
- Taffeta embossing pattern
Specifications
Specifications
Length (Metric) | 45.72 m |
Length (English) | 150 ft. |
For Use With (Application) | Marking hazards, trees, danger areas, flagging boundaries and survey markers |
Type | Flagging Tape |
Material | PVC |
Color | Pink |
Adhesive Type | Non-adhesive |
Tensile Strength | 2620 to 2990 psi |
Width (Metric) | 3 cm |
Thickness | 4 mil |
Show More |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction