Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harris Industries Pavement Marking Tapes

Catalog No. 189991131
Encompass
Click to view available options
Color:
White
Yellow

Catalog No. 18-999-1131

Supplier: Harris Industries PT 3WT

Only null left
Add to Cart
Add to Cart

Specifications

Description Heavy Duty
Adhesive Type Pressure-sensitive
Color White
Format Tape
Product Type Pavement Marking Tape
For Use With (Application) Construction sites and permanently marking driveways and parking lots
Quantity 1 Ea.
Length (English) 50 yd.
Length (Metric) 45.7 m
Width (English) 4 in.
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.