Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HARS Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501979963
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9963 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9963 Supplier Bioss Supplier No. BS20281R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. The protein encoded by this gene is a cytoplasmic enzyme which belongs to the class II family of aminoacyl-tRNA synthetases. The enzyme is responsible for the synthesis of histidyl-transfer RNA, which is essential for the incorporation of histidine into proteins. The gene is located in a head-to-head orientation with HARSL on chromosome five, where the homologous genes share a bidirectional promoter. The gene product is a frequent target of autoantibodies in the human autoimmune disease polymyositis/dermatomyositis.

Specifications

Antigen HARS
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 15mM sodium azide; pH
Gene HARS1
Gene Accession No. P12081
Gene Alias BOS_8256; CMT2W; dead end homolog 1; Dnd1; FLJ20491; Hars; HARSL; HisRS; histidine translase; histidine tRNA ligase; Histidine--tRNA ligase, cytoplasmic; Histidyl-tRNA synthetase; histidyl-tRNA synthetase, cytoplasmic; HRS; MMHRS; USH3B
Gene Symbols HARS1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HARS.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3035
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.