Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HARS2 Antibody (OTI5H2), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271739B
Description
HARS2 Monoclonal specifically detects HARS2 in Human, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
HARS2 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
HARS2 | |
Full length recombinant protein of human HARS2 (NP_036340) produced in HEK293T cells. | |
0.1 mL | |
23438.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI5H2 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
EC 6.1.1, EC 6.1.1.21, HARSLprobable histidyl-tRNA synthetase, mitochondrial, HARS-related, HARSRhistidine tRNA ligase 2, mitochondrial (putative), HisRS, histidine translase, Histidine--tRNA ligase, Histidine--tRNA ligase-like, histidyl-tRNA synthetase 2, mitochondrial (putative), histidyl-tRNA synthetase-like, HO3histidine-tRNA ligase homolog | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Canine, Primate, Monkey | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction