Learn More
Invitrogen™ HARS2 Monoclonal Antibody (OTI2D6)

Description
HARS2 Monoclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | HARS2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI2D6 |
| Concentration | 0.36 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | HARS2 |
| Gene Accession No. | P49590 |
| Gene Alias | 4631412B19Rik; D-tyrosyl-tRNA deacylase 1 homolog; EC 6.1.1.21; EGK_16929; HARS2; Hars2l; HARSL; HARSR; HARS-related; HisRS; histidine translase; histidine tRNA ligase 2, mitochondrial (putative); histidine-tRNA ligase homolog; Histidine--tRNA ligase-like; Histidyl-tRNA synthetase; histidyl-tRNA synthetase 2; histidyl-tRNA synthetase 2, mitochondrial; histidyl-tRNA synthetase 2, mitochondrial (putative); histidyl-tRNA synthetase 2, mitochondrial, hisRS; histidyl-tRNA synthetase-like; HO3; PRLTS2; Probable histidine--tRNA ligase, mitochondrial; probable histidyl-tRNA synthetase, mitochondrial; probable histidyl-tRNA synthetase, mitochondrial-like; putative histidyl-tRNA synthetase, mitochondrial; RGD1308426; zinc finger, matrin type 2; Zmat2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.