Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HARS2 Monoclonal Antibody (OTI5H2)
GREENER_CHOICE

Catalog No. PIMA525754
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525754 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525754 Supplier Invitrogen™ Supplier No. MA525754
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HARS2 Monoclonal Antibody for Western Blot, IHC (P), Flow

Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. The protein encoded by this gene is an enzyme belonging to the class II family of aminoacyl-tRNA synthetases. Functioning in the synthesis of histidyl-transfer RNA, the enzyme plays an accessory role in the regulation of protein biosynthesis. The gene is located in a head-to-head orientation with HARS on chromosome five, where the homologous genes likely share a bidirectional promoter. Mutations in this gene are associated with the pathogenesis of Perrault syndrome, which involves ovarian dysgenesis and sensorineural hearing loss. Alternative splicing results in multiple transcript variants of this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HARS2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5H2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HARS2
Gene Accession No. P49590
Gene Alias 4631412B19Rik; D-tyrosyl-tRNA deacylase 1 homolog; EC 6.1.1.21; EGK_16929; HARS2; Hars2l; HARSL; HARSR; HARS-related; HisRS; histidine translase; histidine tRNA ligase 2, mitochondrial (putative); histidine-tRNA ligase homolog; Histidine--tRNA ligase-like; Histidyl-tRNA synthetase; histidyl-tRNA synthetase 2; histidyl-tRNA synthetase 2, mitochondrial; histidyl-tRNA synthetase 2, mitochondrial (putative); histidyl-tRNA synthetase 2, mitochondrial, hisRS; histidyl-tRNA synthetase-like; HO3; PRLTS2; Probable histidine--tRNA ligase, mitochondrial; probable histidyl-tRNA synthetase, mitochondrial; probable histidyl-tRNA synthetase, mitochondrial-like; putative histidyl-tRNA synthetase, mitochondrial; RGD1308426; zinc finger, matrin type 2; Zmat2
Gene Symbols HARS2
Host Species Mouse
Immunogen Full length human recombinant protein of HARS2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23438, 478035
Target Species Human, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.