Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HARS2 Mouse anti-Human, Clone: OTI6F9, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA503866S

Description
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. The protein encoded by this gene is an enzyme belonging to the class II family of aminoacyl-tRNA synthetases. Functioning in the synthesis of histidyl-transfer RNA, the enzyme plays an accessory role in the regulation of protein biosynthesis. The gene is located in a head-to-head orientation with HARS on chromosome five, where the homologous genes share a bidirectional promoter.Specifications
HARS2 | |
Monoclonal | |
0.6 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P49590 | |
HARS2 | |
Full length human recombit protein of human HARS2 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI6F9 | |
Unconjugated | |
HARS2 | |
HARSL, HARSR, HO3, PRLTS2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23438 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction