Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTX™ Needle L-Shaped Platinum Electrode Kit for Electroporators

Catalog No. btx450510 Shop All BTX Harvard Products
Click to view available options
Pulse Form:
Square Wave

Needle L-Shaped Platinum Electrode Kit with ultra thin diameter electrodes for in vivo applications.

  • Accessory for Electroporators
  • Enables pinpoint transfection for greater ease and efficiency in fragile tissue
  • L-shaped electrodes available in 3 mm tip length

Specifications

Format Kit
For Use With (Application) Ex-vivo or in-vivo tissues gene or drug delivery. Nuclear Transfer
For Use With (Equipment) ECM830, Gemini X2, ECM2001+
Material Platinum
Product Line BTX Electroporation
Product Type L-shaped Electrodes
Voltage 100 V
Electrode Shape L-shaped
Includes 2 extended L-shaped platinum needles ultra thin electrodes and connecting cable
Length (English) Electrode 0.12 in.
Length (Metric) Electrode 3 mm
Quantity 1 Ea.
Sterility Non-sterile
Connection Type Banana Plug to Micrograbber
Pulse Form Square Wave
Show More Show Less

Intended for Research use only. Not for use in diagnostic or clinical procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.