Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HAS1 Monoclonal Antibody (5B5B4)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA548486
Description
HAS1 Monoclonal Antibody for Western Blot, Flow, ELISA
Hyaluronic acid is a high molecular weight unbranched polysaccharide synthesized by a wide variety of organisms from bacteria to mammals, and is a constituent of the extracellular matrix. It consists of alternating glucuronic acid and N-acetylglucosamine residues that are linked by beta-1-3 and beta-1-4 glycosidic bonds. HA is synthesized by membrane-bound synthase at the inner surface of the plasma membrane, and the chains are extruded through pore-like structures into the extracellular space. It serves a variety of functions, including space filling, lubrication of joints, and provision of a matrix through which cells can migrate. HA is actively produced during wound healing and tissue repair to provide a framework for ingrowth of blood vessels and fibroblasts. Changes in the serum concentration of HA are associated with inflammatory and degenerative arthropathies such as rheumatoid arthritis. In addition, the interaction of HA with the leukocyte receptor CD44 is important in tissue-specific homing by leukocytes, and overexpression of HA receptors has been correlated with tumor metastasis. HAS1 is a member of the newly identified vertebrate gene family encoding putative hyaluronan synthases, and its amino acid sequence shows significant homology to the hasA gene product of Streptococcus pyogenes.
Specifications
HAS1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q92839 | |
Has1 | |
Purified recombinant fragment of human HAS1 (AA: (74-399)) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
ELISA, Flow Cytometry, Western Blot, Immunohistochemistry, Western Blot | |
5B5B4 | |
Unconjugated | |
Has1 | |
HA synthase 1; HAS; Has1; huHAS1; Hyaluronan synthase 1; hyaluronan synthase1; hyaluronate synthase 1; Hyaluronic acid synthase 1 | |
Mouse | |
Protein G | |
RUO | |
3036 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction