Learn More
Invitrogen™ HAS3 Monoclonal Antibody (3C9)

Description
MA5-17088 targets HAS3 in FACS, IHC, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17088 immunogen is purified recombinant fragment of human HAS3 expressed in E. Coli. MA5-17088 detects HAS3 which has a predicted molecular weight of approximately 63kDa.
Specifications
Specifications
| Antigen | HAS3 |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 3C9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | HAS3 |
| Gene Accession No. | O00219 |
| Gene Alias | HA synthase 3; Has3; Hyaluronan synthase 3; hyaluronate synthase 3; Hyaluronic acid synthase 3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.