Learn More
Haspin Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$155.13 - $588.46
Specifications
Antigen | Haspin |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2510
![]() |
Bethyl Laboratories, Inc
IHC00671T |
10 μL |
Each for $155.13
|
|
|||||
50-157-2509
![]() |
Bethyl Laboratories, Inc
IHC00671 |
100 μL |
Each for $588.46
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Gorilla
GSG2 is required for normal alignment of chromosomes at metaphase.Specifications
Haspin | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
germ cell associated 2 (haspin), germ cell-specific gene 2 protein, haploid germ cell-specific nuclear protein kinase, haspin, H-haspin | |
HASPIN | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q8TF76 | |
83903 | |
Between 300 and 350 | |
Primary | |
4° C | |
HASPIN |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.