Learn More
HAT1/KAT1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | HAT1/KAT1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-0227
![]() |
Bethyl Laboratories, Inc
A305360AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat
Histone acetylation, particularly of histone H4, has been proposed to play an important role in replication-dependent nucleosome assembly. The HAT1 protein contains D, A, and B motifs, which are present in many N-acetyltransferases, including those that acetylate substrates other than histones. The HAT1 holoenzyme consists of 2 subunits: the catalytic 46-kD HAT1 and the accessory p46. The p46 subunit stimulated the activity of HAT1 and bound to core histones. The HAT1 holoenzyme acetylated newly synthesized but not nucleosomal histone H4 at lys5 and lys12, and, to a lesser extent, histone H2A at lys5. HAT1 and p46 polypeptides are located in the nucleus of S-phase cells. HAT1 may play a role in telomeric silencing.Specifications
HAT1/KAT1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
KAT1 | |
Between 369 and 419 | |
Primary | |
4° C | |
HAT1 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O14929, Q8BY71 | |
107435, 8520 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.